Learn More
Rabbit anti-Pig IgG F(ab')2 fragment,Polyclonal Secondary Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | Rabbit anti-Pig IgG F(ab')2 fragment |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit anti-pig IgG recognizes pig IgG F(ab')2 fragment. This secondary antibody was purified using delipidation and defibrination. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized from 10mM sodium phosphate, 250mM NaCl, pH 7.4 (0.01% (w/v) sodium azide) |
| Host Species | Rabbit |
| Immunogen | Pig IgG F(ab')2 fragment |
| Purification Method | This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against pig IgG F(ab')2 and pig serum. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.