Learn More
Rabbit anti-Pig IgM mu heavy chain,Polyclonal Secondary Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | Rabbit anti-Pig IgM mu heavy chain |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit anti-pig IgG recognizes pig IgM mu heavy chain. This secondary antibody was purified using delipidation and defibrination. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized from 20mM potassium phosphate, 150mM NaCl, pH 7.2 (0.01% (w/v) sodium azide) |
| Host Species | Rabbit |
| Immunogen | Pig IgM mu heavy chain |
| Purification Method | This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against pig IgM and pig serum. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.