Learn More
Immunology Consultants anti-Pig Whole Serum Antibody (Rabbit) - Antiserum
SDP
Catalog No. 502248970
Encompass_Preferred
This rabbit antiserum provides broad reactivity to porcine serum proteins, supporting reliable detection in immunoassays, veterinary research, and diagnostic development. Supplied at 1 ml, it delivers strong, multi‑epitope binding suitable for ELISA, immunodiffusion, and general antibody‑based assays. Manufactured by Immunology Consultants, the reagent offers consistent performance and dependable recognition of pig serum components.
This item is not returnable. View return policy
Catalog No. 50-224-8970 Supplier Immunology Consultants Supplier No. RWS5
This rabbit antiserum provides broad reactivity to porcine serum proteins, supporting reliable detection in immunoassays, veterinary research, and diagnostic development. Supplied at 1 ml, it delivers strong, multi‑epitope binding suitable for ELISA, immunodiffusion, and general antibody‑based assays. Manufactured by Immunology Consultants, the reagent offers consistent performance and dependable recognition of pig serum components.