Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Pigeon IgG (H+L) Secondary Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NBP269423
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-69423 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-69423 Supplier R&D Systems Supplier No. NBP269423

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Pigeon samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate FITC
Dilution ELISA 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Formulation PBS (pH 7.2) with No Preservative
Host Species Rabbit
Immunogen Purified normal IgG isolated from pooled pigeon serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 2 mL
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity The reactivity of the antiserum is directed to the Fc and Fab subunits of the IgG molecule. It includes a certain degree of reactivity with other immunoglobulins via the common Fab portion. It does not react with any non-Ig protein in pigeon serum, as tested by immunoelectrophoresis and double radial immunodiffusion.
Reconstitution It is reconstituted by adding 2 ml sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below - 20C. Prior to use, an aliquot is thawed slowly in the dark at ambient temperature, spun down
Target Species Pigeon
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.