Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rabbit Anti-Rat IgG H&L (HRP) | 100 UL
SDP

Catalog No. 5000443676
Encompass_Preferred

This HRP-conjugated Rabbit Anti-Rat IgG H&L antibody is provided as a liquid at a concentration of 1.0 mg/mL. It is supplied in 0.01M Phosphate Buffered Saline, pH 7.2, with 1% BSA, 50% glycerol, and 0.05% Sodium Azide. This antibody is designed for research use only.

  • HRP-conjugated for detection
  • Reactive with rat IgG
  • Suitable for Western blot, IHC-P, and ELISA applications
  • Recommended dilutions for WB: 1/5000-1/20000
  • Recommended dilutions for IHC: 1/500-1/1000
  • Recommended dilutions for ELISA: 1/5000-1/20000
  • Stable for 1 year when stored at -20°C
  • Avoid repeated freeze/thaw cycles
  • Protect from light

Catalog No. 50-004-43676 Supplier Medchemexpress LLC Supplier No. HYP83738100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This HRP-conjugated Rabbit Anti-Rat IgG H&L antibody is provided as a liquid at a concentration of 1.0 mg/mL. It is supplied in 0.01M Phosphate Buffered Saline, pH 7.2, with 1% BSA, 50% glycerol, and 0.05% Sodium Azide. This antibody is designed for research use only.

  • HRP-conjugated for detection
  • Reactive with rat IgG
  • Suitable for Western blot, IHC-P, and ELISA applications
  • Recommended dilutions for WB: 1/5000-1/20000
  • Recommended dilutions for IHC: 1/500-1/1000
  • Recommended dilutions for ELISA: 1/5000-1/20000
  • Stable for 1 year when stored at -20°C
  • Avoid repeated freeze/thaw cycles
  • Protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.