Learn More
Rabbit anti-Sheep IgG (H+L) Secondary Antibody, Alkaline Phosphatase, R&D Systems™

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Dot Blot, ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alkaline Phosphatase (AP) |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:5000, Immunohistochemistry 1:200 - 1:1000, Dot Blot |
| Host Species | Rabbit |
| Immunogen | Rabbit anti-Sheep IgG (H+L) Secondary Antibody [Alkaline Phosphatase] was produced by repeated immunization with Sheep IgG whole molecule in rabbit. |
| Purification Method | Multi-step |
| Quantity | 0.1 mg |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.