Learn More
Immunology Consultants anti-Sheep IgG h+l Antibody (Rabbit) - Affinity Purified
SDP
Catalog No. 502248888
Encompass_Preferred
RGHL‑35A is an affinity‑purified rabbit antibody specific for sheep IgG heavy and light chains. This reagent provides strong, selective binding and low background, making it ideal for use as a secondary antibody in ELISA, Western blotting, and immunodetection workflows involving sheep primary antibodies. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑35A delivers consistent, high‑quality performance for laboratories working with ovine immunoglobulins.
This item is not returnable. View return policy
Catalog No. 50-224-8888 Supplier Immunology Consultants Supplier No. RGHL35A
RGHL‑35A is an affinity‑purified rabbit antibody specific for sheep IgG heavy and light chains. This reagent provides strong, selective binding and low background, making it ideal for use as a secondary antibody in ELISA, Western blotting, and immunodetection workflows involving sheep primary antibodies. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑35A delivers consistent, high‑quality performance for laboratories working with ovine immunoglobulins.