Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Rabbit anti-Sheep, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. 12342MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-342-MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-342-MI Supplier MilliporeSigma Supplier No. 12342
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Rabbit anti-Sheep IgG Antibody, HRP conjugate detects level of Sheep IgG and has been published and validated for use in ELISA, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabbit anti-Sheep IgG
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation 0.02M Potassium Phosphate, 0.15M NaCl, pH 7.2, 10mg/mL BSA, and 0.01% gentamicin sulfate
Host Species Rabbit
Quantity 500 μg
Primary or Secondary Secondary
Target Species Sheep
Content And Storage Lyophilized: 2 years at 4°C; Rehydrated: 6 months at −20°C.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.