Learn More
Innovative Research Inc Rabbit Anti Sheep IgG Polyclonal Affinity Purified
Catalog No. NC2660024
Encompass_Preferred
Rabbit Anti Sheep IgG Polyclonal Affinity Purified from Innovative Research is a polyclonal antibody in Liquid format, buffered in Phospate buffered saline, pH 7.2 with 0.1% sodium azide. Rabbits were immunized with purified Sheep IgG in Freund’s adjuvant. Anti IgG was immunoaffinity purified using antigen coupled to agarose beads.
Catalog No. NC2660024 Supplier Innovative Research Inc Supplier No. IRBASHIGGAP1MG
Rabbit Anti Sheep IgG Polyclonal Affinity Purified from Innovative Research is a polyclonal antibody in Liquid format, buffered in Phospate buffered saline, pH 7.2 with 0.1% sodium azide. Rabbits were immunized with purified Sheep IgG in Freund’s adjuvant. Anti IgG was immunoaffinity purified using antigen coupled to agarose beads.