Learn More
Cayman Chemical Rabbit Anti-Sheep IgG Precoat
SDP
Catalog No. 502942434
Encompass_Preferred
An MTTP inhibitor (IC50 2 nM) inhibits ApoB and triglyceride secretion in HepG2 cells (IC50 2.6 nM for both) lowers plasma triglyceride levels as well as plasma total VLDL and LDL cholesterol levels in rats at 10 mg/kg
Catalog No. 50-294-2434 Supplier Cayman Chemical Supplier No. 4006585 EA
An MTTP inhibitor (IC50 2 nM) inhibits ApoB and triglyceride secretion in HepG2 cells (IC50 2.6 nM for both) lowers plasma triglyceride levels as well as plasma total VLDL and LDL cholesterol levels in rats at 10 mg/kg