Learn More
Rabbit anti-Sheep IgG, Whole Molecule, FITC, Polyclonal Secondary Antibody, MP Biomedicals
Shop All MP Biomedicals, Inc ProductsDescription
Host Species: Rabbit
Species Reactivity: Sh
Immunofluorescence, Fluorescence Microscopy, Fluorescent Activated Cell Sorting, Western Blotting
Specifications
Specifications
| Antigen | IgG |
| Classification | Polyclonal |
| Conjugate | FITC |
| Host Species | Rabbit |
| Purification Method | Affinity Purified |
| Quantity | 2 mg |
| Regulatory Status | RUO |
| Primary or Secondary | Secondary |
| Target Species | Sheep |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.