Learn More
Immunology Consultants anti-Sheep Whole Serum Antibody (Rabbit) - Antiserum
SDP
Catalog No. 502248972
Encompass_Preferred
This rabbit antiserum against sheep whole serum provides comprehensive recognition of ovine serum proteins, making it suitable for ELISA, immunodiffusion, and assay development. Produced by Immunology Consultants, this 1 ml reagent offers strong, multi‑epitope binding and consistent performance across research and diagnostic workflows. Its broad reactivity supports robust detection of sheep serum components in diverse sample types.
This item is not returnable. View return policy
Catalog No. 50-224-8972 Supplier Immunology Consultants Supplier No. RWS35
This rabbit antiserum against sheep whole serum provides comprehensive recognition of ovine serum proteins, making it suitable for ELISA, immunodiffusion, and assay development. Produced by Immunology Consultants, this 1 ml reagent offers strong, multi‑epitope binding and consistent performance across research and diagnostic workflows. Its broad reactivity supports robust detection of sheep serum components in diverse sample types.