Learn More
Seracare Life Sciences Inc RABBIT ANTI-SHIGELLA SPECIES
Supplier:  Seracare Life Sciences Inc 53300066
Encompass_Preferred
Protein G purified BacTrace antibody to Shigella species labeled with DyLight 488. Isolated from a serum pool of rabbits immunized with heat-killed whole cells of various strains of Shigella.
Catalog No. 50-670-93