Learn More
Seracare Life Sciences Inc RABBIT ANTI-VIBRIO SPECIES
Supplier:  Seracare Life Sciences Inc 53300067
Encompass_Preferred
Protein G purified BacTrace antibody to Vibrio species labeled with DyLight 488. Isolated from a serum pool of rabbits immunized with heat-killed whole cells of various strains of Vibrio.
Catalog No. 50-670-94