Learn More
Seracare Life Sciences Inc RABBIT ANTI-YERSINIA SPECIES
Supplier:  Seracare Life Sciences Inc 53300069
Encompass_Preferred
Protein G purified BacTrace antibody to Yersinia species labeled with DyLight 488. Isolated from a serum pool of rabbits immunized with various strains of heat-killed Yersinia.
Catalog No. 50-670-96