Learn More
Seracare Life Sciences Inc RABBIT ANTI-YERSINIA SPECIES
Supplier:  Seracare Life Sciences Inc 53100313
Encompass_Preferred
Protein G purified BacTrace antibody isolated from a serum pool of rabbits immunized with various strains of heat-killed Yersinia.
Catalog No. 50-668-55