Learn More
Aviva Systems Biology Corp exd Antibody - C-terminal region (ARP47897_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47897_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against exd. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Fruit fly. Alternate names: Dmel_CG8933, CG8933, DExd, Dm-EXD, Dpbx, EXD, td48, unnamed, anon-EST:fe1H3, Dmel\CG8933, Exd, l(1)IV, Pbx1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59680