Learn More
Aviva Systems Biology Corp lab Antibody - N-terminal region (ARP47876_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47876_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against lab. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide corresponding to a region of Fruit fly. Alternate names: Dmel_CG1264, BG:DS00004.9, CG1264, DmLab, EfR9, F121, F24, F90, F90-2, l(3)01241, l(3)84Ac, lb, Dm lab, Dmel\CG1264, Dmlab, LabApplicationWB. Species reactivity:
Catalog No. 50-014-59677