Learn More
Aviva Systems Biology Corp cad Antibody - middle region (ARP47825_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47825_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against cad. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Fruit fly. Alternate names: Dmel_CG1759, 38E.19, CG1759, Cad, S67, cd, anon-WO2004063362.83, CAD, Dmel\CG1759ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59673