Learn More
Aviva Systems Biology Corp eve Antibody - N-terminal region (ARP47848_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47848_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against eve. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Fruit fly. Alternate names: Dmel_CG2328, 10.5, 10.9, 14.10, 20.35, CG2328, E(eve), EVE, F, V, VI, eve2, l(2)46Ce, Dmel\CG2328, Eve, l(2)46CFg, l(2)46CFh, l(2)46CFj, l(2)46CFp, l(2)46CgApplicationWB. Species reactivity:
Catalog No. 50-014-59675