Learn More
Cayman Chemical Rabbit AntiMouse IgA Polyclon
SDP
Catalog No. 502886652
Encompass_Preferred
Antigen: mouse IgA · Host: rabbit · Cross Reactivity: (+) mouse IgA; (−) murine IgG · Applications: FC and IF
Catalog No. 50-288-6652 Supplier Cayman Chemical Supplier No. 101961 EA
Antigen: mouse IgA · Host: rabbit · Cross Reactivity: (+) mouse IgA; (−) murine IgG · Applications: FC and IF