Learn More
Rabbit antiserum to Human Immunoglobulins Polyclonal Secondary Antibody, MP Biomedicals
Shop All MP Biomedicals, Inc ProductsDescription
Host Species: Rabbit
Species Reactivity: Hu
Immunoelectrophoresis, Immunodiffusion, Immunohistochemistry (Paraffin/Frozen)
Specifications
Specifications
| Antigen | IgG |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 2 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Secondary |
| Target Species | Human |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.