Learn More
American Research Products Inc Rabbit apolipoprotein A1 Apo
Catalog No. 5000326172
Encompass
Rabbit apolipoprotein A1 (Apo-A1) ELISA Kit from CUSABIO Detection Range 6 25 g/mL-400 g/mL Sensitivity 1 56 g/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26172 Supplier American Research Products Inc Supplier No. CSBE09804RB
Rabbit apolipoprotein A1 (Apo-A1) ELISA Kit from CUSABIO Detection Range 6 25 g/mL-400 g/mL Sensitivity 1 56 g/mL Sample serum plasma tissue homogenates Method Sandwich