Learn More
Biomatik Corporation Rabbit Apolipoprotein E (APOE) ELISA Kit
Supplier:  Biomatik Corporation EKU02521
Encompass_Preferred
Rabbit Apolipoprotein E (APOE) ELISA Kit for detecting APOE in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 493.8-40,000pg/ml. Sensitivity: 182.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P18287. Gene ID: 100009337. Target Synonyms: Apo-E; AD2; Apoprotein; Alzheimer Disease 2 (E4-Associated,Late Onset. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9124