Learn More
Biomatik Corporation Rabbit Aquaporin 2, Collecting Duct (AQP2) ELISA Kit
Supplier:  Biomatik Corporation EKU11913
Encompass_Preferred
Rabbit Aquaporin 2, Collecting Duct (AQP2) ELISA Kit for detecting AQP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A5F9CSK7. Target Synonyms: AQP-CD; WCH-CD; ADH water channel; Aquaporin-CD; Collecting duct water channel protein; Water channel protein for renal collecting duct. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8066