Learn More
American Research Products Inc Rabbit arachidonic Acid AA E
Catalog No. 5000328054
Encompass
Rabbit arachidonic Acid (AA) ELISA kit from CUSABIO Detection Range 78 125 ng/mL-5000 ng/mL Sensitivity 69 748 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28054 Supplier American Research Products Inc Supplier No. CSBEQ027590RB
Rabbit arachidonic Acid (AA) ELISA kit from CUSABIO Detection Range 78 125 ng/mL-5000 ng/mL Sensitivity 69 748 ng/mL Sample serum plasma tissue homogenates Method Competitive