Learn More
American Research Products Inc Rabbit b Thromboglobulin b TG
Catalog No. 5000328717
Encompass
Rabbit b-Thromboglobulin b-TG ELISA Kit from CUSABIO Detection Range 12 5 ng/mL-800 ng/mL Sensitivity 3 12 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28717 Supplier American Research Products Inc Supplier No. CSBE09285RB
Rabbit b-Thromboglobulin b-TG ELISA Kit from CUSABIO Detection Range 12 5 ng/mL-800 ng/mL Sensitivity 3 12 ng/mL Sample serum plasma tissue homogenates Method Sandwich