Learn More
Kingfisher Biotech Inc Rabbit BAFF Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0927U025
Encompass
The Rabbit BAFF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Rabbit BAFF applications are for cell culture, ELISA standard, and Western Blot Control. The Rabbit BAFF yeast-derived recombinant protein can be purchased in multiple sizes. Rabbit BAFF Specifications: (Molecular Weight: 17.1 kDa) (Amino Acid Sequence: AVEGVEETVI QDCLQLIADS DTPIIRKGSY TFVPWLLSFK RGRALEEKEN KIVVKETGYF FIYGQVLYTD STFAMGHLIQ RKKVHVFGDE LSLVTLFRCI QNMPETLPNN SCYSAGIAKL EEGDELQLAI PRENAQISRD GDGTFFGALK LL (152)) (Gene ID: 100101577). For research use only. Made in the USA
Catalog No. 50-241-3971