Learn More
American Research Products Inc Rabbit basic fibroblast growth
Catalog No. 5000329824
Encompass
Rabbit basic fibroblast growth factor bFGF ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29824 Supplier American Research Products Inc Supplier No. CSBE06938RB
Rabbit basic fibroblast growth factor bFGF ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich