Learn More
American Research Products Inc Rabbit Bone Alkaline Phosphata
Catalog No. 5000328633
Encompass
Rabbit Bone Alkaline Phosphatase (BALP) ELISA Kit from CUSABIO Detection Range 0 156 mIU/mL-10 mIU/mL Sensitivity 0 039 mIU/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-28633 Supplier American Research Products Inc Supplier No. CSBE15042RB
Rabbit Bone Alkaline Phosphatase (BALP) ELISA Kit from CUSABIO Detection Range 0 156 mIU/mL-10 mIU/mL Sensitivity 0 039 mIU/mL Sample serum plasma Method Sandwich