Learn More
American Research Products Inc Rabbit C ReactiVe Protein CRP
Catalog No. 5000326449
Encompass
Rabbit C-ReactiVe Protein CRP ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-2000 pg/mL Sensitivity 2 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-26449 Supplier American Research Products Inc Supplier No. CSBE06847RB
Rabbit C-ReactiVe Protein CRP ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-2000 pg/mL Sensitivity 2 pg/mL Sample serum plasma Method Sandwich