Learn More
Kingfisher Biotech Inc Chicken TNFSF15 ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1220U004
Encompass_Preferred
The Chicken TNFSF15 (TL1A, VEGI) ELISA contains capture antibody, standard, and detection antibody for development of a Chicken TNFSF15 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include TNFSF15, TL1, TL1A, VEGI, VEGI192A, Vascular endothelial growth inhibitor, TNLG1B, tumor necrosis factor superfamily member 15, TNF superfamily member 15. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5372