Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabbit C3a ELISA Kit

Catalog No. ER3RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ER3RBX5 5 x 96 Tests
ER3RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ER3RBX5 Supplier Invitrogen™ Supplier No. ER3RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Rabbit C3a quantitates rabbit C3a in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rabbit C3a.

Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12247
Assay Range 0.1-20 ng/mL
Assay Sensitivity 0.1 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Rabbit
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3r1; Complement C3; complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3 alpha-chain; complement component C3a; complement component C3b; CPAMD1; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; Plp; prepro-C3Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3r1; Complement C3; complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3 alpha-chain; complement component C3a; complement component C3b; CPAMD1; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; Plp; prepro-C3;
Gene ID (Entrez) 100009121
Gene Symbol C3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Complement C3a
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.