Learn More
American Research Products Inc Rabbit cardiac troponin I cTn
Catalog No. 5000326220
Encompass
Rabbit cardiac troponin cTn- ELISA Kit from CUSABIO Detection Range 47 pg/mL-3000 pg/mL Sensitivity 11 75 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26220 Supplier American Research Products Inc Supplier No. CSBE11286RB
Rabbit cardiac troponin cTn- ELISA Kit from CUSABIO Detection Range 47 pg/mL-3000 pg/mL Sensitivity 11 75 pg/mL Sample serum plasma tissue homogenates Method Sandwich