Learn More
Kingfisher Biotech Inc Rabbit CCL2 Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0431U100
Encompass
The Rabbit CCL2 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Rabbit CCL2 applications are for cell culture, ELISA standard, and Western Blot Control. The Rabbit CCL2 yeast-derived recombinant protein can be purchased in multiple sizes. Rabbit CCL2 Specifications: (Molecular Weight: 11.4 kDa) (Amino Acid Sequence: QPDAVNSPVT CCYTFTNKTI SVKRLMSYRR INSTKCPKEA VIFMTKLAKG ICADPKQKWV QDAIANLDKK MQTPKTLTSY STTQEHTTNL SSTRTPSTTT SL (102)) (Gene ID: 100009130). For research use only. Made in the USA
Catalog No. 50-241-3296