Learn More
ACROBiosystems Mouse Anti-RSV-F0 Antibody IgG Titer ELISA Assay Kit (Pre-Fusion glycoprotein F0)
Catalog No. 502718988
Encompass_Preferred
Respiratory syncytial virus (RSV) is a highly contagious virus causing severe infection in infants and the elderly. Various approaches are being used to develop an effective RSV vaccine. The RSV fusion (F) subunit, particularly the cleaved trimeric pre-fusion F, is one of the most promising vaccine candidates under development.To facilitate the RSV-related research, drug trials and vaccine development, a high-throughput assay to measure IgG antibodies against the virus is in urgent need.
Catalog No. 50-271-8988 Supplier ACROBiosystems Supplier No. RAST17496TESTS
Respiratory syncytial virus (RSV) is a highly contagious virus causing severe infection in infants and the elderly. Various approaches are being used to develop an effective RSV vaccine. The RSV fusion (F) subunit, particularly the cleaved trimeric pre-fusion F, is one of the most promising vaccine candidates under development.To facilitate the RSV-related research, drug trials and vaccine development, a high-throughput assay to measure IgG antibodies against the virus is in urgent need.