Learn More
ACROBiosystems Rabbit CD3E&CD3D Heterodimer Protein, Llama Fc&Llama Fc, low endotoxin (MALS verified)
Supplier:  ACROBiosystems CDDR5259100UG
Encompass_Preferred
Recombinant Protein;100UG;Rabbit CD3E&CD3D Heterodimer Protein, Llama Fc&Llama Fc (CDD-R5259) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Thr 120 (CD3E) & Thr 22 - Ser 107 (CD3D) (Accession # Q9TUF9-1 (CD3E) & A0A5F9D3I4-1 (CD3D)).;Rabbit CD3E&CD3D Heterodimer Protein, Llama Fc&Llama Fc is produced by co-expression of CD3E and CD3D, has a calculated MW of 42.4 kDa (CD3E) and 40.8 kDa (CD3D). Subunit CD3E is fused with a llama IgG2b Fc tag at the C-terminus and subunit CD3D is fused with a llama IgG2b Fc tag at the C-terminus. The reducing (R) protein migrates as 50 kDa and 60 kDa due to glycosylation.
Catalog No. 50-205-4838