Learn More
ACROBiosystems Rabbit CD3E&CD3D Heterodimer Protein, Llama Fc Tag, Low Endotoxin, 1 mg
Catalog No. 502054837
Encompass_Preferred
This Rabbit CD3E and CD3D heterodimer protein consists of the T-cell surface glycoprotein CD3 delta and CD3 epsilon chains, which are essential components of the T cell receptor-CD3 complex involved in coupling antigen recognition to intracellular signal-transduction pathways. This protein is expressed in HEK293 cells and features a llama Fc tag on both subunits.
  • Expressed in human 293 cells (HEK293)
  • Purity >95% as determined by SDS-PAGE
  • Low endotoxin level of less than 0.01 EU per μg
  • Lyophilized from a 0.22 μm filtered solution in PBS, pH 7.4 with trehalose
  • Calculated molecular weight of 42.4 kDa (CD3E) and 40.8 kDa (CD3D)
  • Validated by MALS
Catalog No. 50-205-4837 Supplier ACROBiosystems Supplier No. CDDR52591MG
This Rabbit CD3E and CD3D heterodimer protein consists of the T-cell surface glycoprotein CD3 delta and CD3 epsilon chains, which are essential components of the T cell receptor-CD3 complex involved in coupling antigen recognition to intracellular signal-transduction pathways. This protein is expressed in HEK293 cells and features a llama Fc tag on both subunits.
  • Expressed in human 293 cells (HEK293)
  • Purity >95% as determined by SDS-PAGE
  • Low endotoxin level of less than 0.01 EU per μg
  • Lyophilized from a 0.22 μm filtered solution in PBS, pH 7.4 with trehalose
  • Calculated molecular weight of 42.4 kDa (CD3E) and 40.8 kDa (CD3D)
  • Validated by MALS