Learn More
ACROBiosystems Rabbit CD40 Ligand / TNFSF5 Protein, His,Flag Tag (active trimer) (MALS verified)
Catalog No. 502329137
Encompass_Preferred
Rabbit CD40 Ligand, His,Flag Tag (CD0-R52D9) is expressed from human 293 cells (HEK293). It contains AA Gly 116 - Leu 261 (Accession # G1SKP7-1).
Catalog No. 50-232-9137 Supplier ACROBiosystems Supplier No. CD0R52D9100UG
Rabbit CD40 Ligand, His,Flag Tag (CD0-R52D9) is expressed from human 293 cells (HEK293). It contains AA Gly 116 - Leu 261 (Accession # G1SKP7-1).