Learn More
Kingfisher Biotech Inc Rabbit CD40 Ligand Recombinant Protein (Yeast, Endotoxin - Free) - 500 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0990U500
Encompass
The Rabbit CD40 Ligand yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Rabbit CD40 Ligand applications are for cell culture, ELISA standard, and Western Blot Control. The Rabbit CD40 Ligand yeast-derived recombinant protein can be purchased in multiple sizes. Rabbit CD40 Ligand Specifications: (Molecular Weight: 16.2 kDa) (Amino Acid Sequence: MQKGDQDPQI AAHLISEASS KSSSVLQWAK KGYYTMSNTL VTLENGKQLK VKRQGFYYIY AQVTFCSNQE PSSQAPFIAS LCLKSSGGSE RILLRAANAR SSSKTCEQQS IHLGGVFELQ ADASVFVNVT DASQVNHGTG FTSFGLLKL (149)) (Gene ID: 100358388). For research use only. Made in the USA
Catalog No. 50-241-4134