Learn More
ACROBiosystems ACROBiosystems Rabbit CD40 / TNFRSF5 Protein, His Tag (MALS verified)
Catalog No. 502106494
Encompass_Preferred
Recombinant Protein;100UG;Rabbit CD40, His Tag (CD0-R52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 77 - Arg 249 (Accession # G1SPC6-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.9 kDa. The protein migrates as 28-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rabbit CD40, His Tag (Cat. No. CD0-R52H9) at 5 μg/mL (100 μL/well) can bind Human CD40 Ligand, Fc Tag (active trimer) (MALS verified) (Cat. No. CDL-H5269) with a linear range of 0.2-1 ng/mL (QC tested).;Immobilized Rabbit CD40, His Tag (Cat. No. CD0-R52H9) at 5 μg/mL (100 μL/well) can bind Anti-CD40 Antibody, Human IgG2 with a linear range of 0.2-2 ng/mL (Routinely tested).
Catalog No. 50-210-6494 Supplier ACROBiosystems Supplier No. CD0R52H9100UG
Recombinant Protein;100UG;Rabbit CD40, His Tag (CD0-R52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 77 - Arg 249 (Accession # G1SPC6-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.9 kDa. The protein migrates as 28-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rabbit CD40, His Tag (Cat. No. CD0-R52H9) at 5 μg/mL (100 μL/well) can bind Human CD40 Ligand, Fc Tag (active trimer) (MALS verified) (Cat. No. CDL-H5269) with a linear range of 0.2-1 ng/mL (QC tested).;Immobilized Rabbit CD40, His Tag (Cat. No. CD0-R52H9) at 5 μg/mL (100 μL/well) can bind Anti-CD40 Antibody, Human IgG2 with a linear range of 0.2-2 ng/mL (Routinely tested).