Learn More
ACROBiosystems ACROBiosystems Rabbit CD73 / NT5E Protein, His Tag
Catalog No. 502106813
Encompass_Preferred
Recombinant Protein;1MG;Rabbit CD73, His Tag (CD3-R52H3) is expressed from human 293 cells (HEK293). It contains AA Trp 27 - Ser 550 (Accession # XP_002714603.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.8 kDa.
Catalog No. 50-210-6813 Supplier ACROBiosystems Supplier No. CD3R52H31MG
Recombinant Protein;1MG;Rabbit CD73, His Tag (CD3-R52H3) is expressed from human 293 cells (HEK293). It contains AA Trp 27 - Ser 550 (Accession # XP_002714603.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.8 kDa.