Learn More
Biomatik Corporation Rabbit Cholecystokinin A Receptor (CCKAR) CLIA Kit
Supplier:  Biomatik Corporation EKU09623
Encompass_Preferred
Rabbit Cholecystokinin A Receptor (CCKAR) CLIA Kit for detecting CCKAR in tissue homogenates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 11.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O97772. Gene ID: 100009241. Target Synonyms: CCK-A; CCK1; CCKRA; CCK1R; Cholecystokinin Type 1 Receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8481