Learn More
American Research Products Inc Rabbit cholesteryl ester trans
Catalog No. 5000326792
Encompass
Rabbit cholesteryl ester transfer protein CETP ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26792 Supplier American Research Products Inc Supplier No. CSBE06841RB
Rabbit cholesteryl ester transfer protein CETP ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Competitive