Learn More
Biomatik Corporation Rabbit Cholesteryl Ester Transfer Protein (CETP) ELISA Kit
Supplier:  Biomatik Corporation EKU03153
Encompass_Preferred
Rabbit Cholesteryl Ester Transfer Protein (CETP) ELISA Kit for detecting CETP in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 46.88-3000pg/ml. Sensitivity: 20.53pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P22687. Target Synonyms: LTP; Lipid Transfer Protein; Plasma Lipid Transfer Protein; Cholesterylester Transfer Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4542