Learn More
Biomatik Corporation Rabbit Ciliary Neurotrophic Factor (CNTF) CLIA Kit
Supplier:  Biomatik Corporation EKU09603
Encompass_Preferred
Rabbit Ciliary Neurotrophic Factor (CNTF) CLIA Kit for detecting CNTF in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 4.12-3,000pg/ml. Sensitivity: 1.93pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P14188. Gene ID: 100009117. Target Synonyms: HCNTF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5405