Learn More
Biomatik Corporation Rabbit Cluster Of Differentiation 40 Ligand (CD40L) ELISA Kit
Supplier:  Biomatik Corporation EKU10723
Encompass_Preferred
Rabbit Cluster Of Differentiation 40 Ligand (CD40L) ELISA Kit for detecting CD40L in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G1SKP7. Gene ID: 100358388. Target Synonyms: CD154; TRAP; HIGM1; IGM; IMD3; TBAM; T-BAM; TNFSF5; Gp39; TNF Superfamily Member 5; Hyper-IgM Syndrome; TNF-Related Activation Protein; T-Cell B-Cell Activating Molecule. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8302