Learn More
American Research Products Inc Rabbit Coagulation factor VII
Catalog No. 5000328132
Encompass
Rabbit Coagulation factor VII (F7) ELISA kit from CUSABIO Detection Range 3 9 ng/mL-250 ng/mL Sensitivity 1 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28132 Supplier American Research Products Inc Supplier No. CSBEL007930RB
Rabbit Coagulation factor VII (F7) ELISA kit from CUSABIO Detection Range 3 9 ng/mL-250 ng/mL Sensitivity 1 ng/mL Sample serum plasma tissue homogenates Method Competitive