Learn More
Biomatik Corporation Rabbit Complement Component 3a (C3a) ELISA Kit
Supplier:  Biomatik Corporation EKU03391
Encompass_Preferred
Rabbit Complement Component 3a (C3a) ELISA Kit for detecting C3a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.3ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12247. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4338