Learn More
American Research Products Inc Rabbit Complement fragment 3a
Catalog No. 5000330165
Encompass
Rabbit Complement fragment 3a C3a ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30165 Supplier American Research Products Inc Supplier No. CSBE09283RB
Rabbit Complement fragment 3a C3a ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates Method Sandwich